Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxj2
Entrez Gene ID
60611
Gene Name
forkhead box J2
Gene Aliases
Fhx
Chromosome Location
Chr.6: 122820184 - 122845366 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021899.3 NP_068699.1 9 - 10 2308 80
GenBank mRNA AB221666.1 - 8 - 9 1507 80
AF253052.1 - 9 - 10 2320 80
BC040395.1 - 10 - 11 1938 80

View Full Product Details