The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146287.1 | NP_001139759.1 | 2 - 3 | 1331 | 71 |
NM_022021.2 | NP_071304.2 | 2 - 3 | 1331 | 71 | |
GenBank mRNA | AF133208.1 | - | 2 - 3 | 1330 | 71 |
AF328140.1 | - | 2 - 3 | 1016 | 71 | |
AK156273.1 | - | 2 - 3 | 1387 | 71 | |
AK158396.1 | - | 2 - 3 | 1061 | 71 | |
BC043661.1 | - | 2 - 3 | 533 | 71 | |
BC145142.1 | - | 2 - 3 | 1315 | 71 | |
BC150714.1 | - | 2 - 3 | 1315 | 71 | |
CX213994.1 | - | 2 - 3 | 392 | 71 |