The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025875.2 | NP_080151.2 | 2 - 3 | 162 | 110 |
GenBank mRNA | AK009953.1 | - | 2 - 3 | 157 | 110 |
AK010284.1 | - | 2 - 3 | 150 | 110 | |
BC020086.1 | - | 2 - 3 | 145 | 110 | |
BC058376.1 | - | 2 - 3 | 134 | 110 | |
BY128307.1 | - | 2 - 3 | 159 | 110 |