The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110826.1 | NP_001104296.1 | 3 - 4 | 623 | 142 |
NM_007841.4 | NP_031867.1 | 3 - 4 | 566 | 142 | |
NM_181324.3 | NP_851841.2 | 3 - 4 | 591 | 142 | |
GenBank mRNA | AF038995.1 | - | 2 - 3 | 185 | 142 |
AK036648.1 | - | 3 - 4 | 619 | 142 | |
AK054144.1 | - | 3 - 4 | 585 | 142 | |
AK148483.1 | - | 3 - 4 | 588 | 142 | |
BC021452.1 | - | 2 - 3 | 483 | 142 | |
CD353843.1 | - | 3 - 4 | 554 | 142 |