The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110826.1 | NP_001104296.1 | 11 - 12 | 1534 | 105 |
NM_007841.4 | NP_031867.1 | 11 - 12 | 1477 | 105 | |
NM_181324.3 | NP_851841.2 | 11 - 12 | 1502 | 105 | |
GenBank mRNA | AK148483.1 | - | 11 - 12 | 1499 | 105 |
BC021452.1 | - | 10 - 11 | 1394 | 105 |