The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007853.4 | NP_031879.1 | 2 - 3 | 882 | 76 |
GenBank mRNA | AF466376.1 | - | 2 - 3 | 899 | 76 |
AK002617.1 | - | 2 - 3 | 890 | 76 | |
AK077257.1 | - | 2 - 3 | 883 | 76 | |
AK149811.1 | - | 2 - 3 | 872 | 76 | |
AK150908.1 | - | 2 - 3 | 871 | 76 | |
AK163404.1 | - | 2 - 3 | 872 | 76 | |
BC003751.1 | - | 2 - 3 | 900 | 76 | |
Y08460.1 | - | 2 - 3 | 867 | 76 |