Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drg1
Entrez Gene ID
13494
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
AA408859, AI132520, Nedd3
Chromosome Location
Chr.11: 3249922 - 3266386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007879.1 NP_031905.1 1 - 2 122 70
GenBank mRNA AK044194.1 - 1 - 2 131 70
AK090156.1 - 1 - 2 152 70
AK144438.1 - 1 - 2 124 70
AK155061.1 - 1 - 2 137 70
BC039649.1 - 1 - 2 116 70
D10715.1 - 1 - 2 122 70

View Full Product Details