The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008130.2 | NP_032156.2 | 5 - 6 | 1109 | 82 |
GenBank mRNA | AK144851.1 | - | 5 - 6 | 893 | 82 |
AK158261.1 | - | 5 - 6 | 1109 | 82 | |
BC141135.1 | - | 5 - 6 | 810 | 82 | |
BC145445.1 | - | 5 - 6 | 810 | 82 | |
X95255.1 | - | 5 - 6 | 849 | 82 |