Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gnl1
Entrez Gene ID
14670
Gene Name
guanine nucleotide binding protein-like 1
Gene Aliases
Gna-rs1, Gnal1
Chromosome Location
Chr.17: 35979955 - 35989462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008136.2 NP_032162.2 9 - 10 1495 84
GenBank mRNA AK078187.1 - 9 - 10 1458 84
BC010298.1 - 9 - 10 1450 84
BC013243.1 - 9 - 10 1519 84
BC094391.1 - 7 - 8 932 84
BC110397.1 - 5 - 6 760 84
BC129969.1 - 7 - 8 1027 84
BC129970.1 - 7 - 8 1027 84
X65026.1 - 8 - 9 1137 84

View Full Product Details