Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thumpd3
Entrez Gene ID
14911
Gene Name
THUMP domain containing 3
Gene Aliases
AW556087, Gt(ROSA)26asSor, Gtrosa26as
Chromosome Location
Chr.6: 113046252 - 113068278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008188.2 NP_032214.1 2 - 3 354 99
GenBank mRNA AK017246.1 - 2 - 3 367 99
AK142140.1 - 2 - 3 373 99
BC002024.1 - 2 - 3 356 99
BC012688.1 - 1 - 2 302 99
BC046800.1 - 2 - 3 354 99
U83176.1 - 2 - 3 348 99

View Full Product Details