Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thumpd3
Entrez Gene ID
14911
Gene Name
THUMP domain containing 3
Gene Aliases
AW556087, Gt(ROSA)26asSor, Gtrosa26as
Chromosome Location
Chr.6: 113046252 - 113068278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008188.2 NP_032214.1 7 - 8 1227 98
GenBank mRNA AK142140.1 - 7 - 8 1246 98
BC046800.1 - 7 - 8 1227 98
U83176.1 - 7 - 8 1221 98

View Full Product Details