The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008196.1 | NP_032222.1 | 3 - 4 | 365 | 79 |
GenBank mRNA | AB032200.1 | - | - | 610 | 79 |
BC117981.1 | - | 3 - 4 | 364 | 79 |