Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ido1
Entrez Gene ID
15930
Gene Name
indoleamine 2,3-dioxygenase 1
Gene Aliases
Ido, Indo
Chromosome Location
Chr.8: 24584133 - 24596952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293690.1 NP_001280619.1 6 - 7 716 64
NM_008324.2 NP_032350.1 6 - 7 563 64
GenBank mRNA AK033783.1 - 6 - 7 717 64
BC049931.1 - 6 - 7 554 64
M69109.1 - 6 - 7 620 64

View Full Product Details