The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008342.3 | NP_032368.2 | 1 - 2 | 484 | 85 |
GenBank mRNA | AK011784.1 | - | 2 - 3 | 355 | 85 |
AK011790.1 | - | 1 - 2 | 413 | 85 | |
AK012703.1 | - | 1 - 2 | 474 | 85 | |
BC012724.1 | - | 1 - 2 | 472 | 85 | |
BC054473.1 | - | 1 - 2 | 457 | 85 | |
X81580.1 | - | 1 - 2 | 455 | 85 |