Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrmp
Entrez Gene ID
16970
Gene Name
lymphoid-restricted membrane protein
Gene Aliases
D6Int3, D6Int4, D6Int5, D6Int7, D6Int8, Jaw1
Chromosome Location
Chr.6: 145111760 - 145174928 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281980.1 NP_001268909.1 9 - 10 733 75
NM_001281981.1 NP_001268910.1 10 - 11 774 75
NM_008511.3 NP_032537.3 9 - 10 1005 75
GenBank mRNA AK040180.1 - 9 - 10 1006 75
AK050646.1 - 9 - 10 1443 75
AK079932.1 - 9 - 10 963 75
AK153823.1 - 9 - 10 1003 75
AK153865.1 - 9 - 10 735 75
AK160009.1 - 9 - 10 965 75
AK166635.1 - 7 - 8 454 75
AK168775.1 - 7 - 8 431 75
AK169578.1 - 10 - 11 758 75
AY753324.1 - 8 - 9 568 75
U10484.1 - 9 - 10 711 75

View Full Product Details