Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrp5
Entrez Gene ID
16973
Gene Name
low density lipoprotein receptor-related protein 5
Gene Aliases
BMND1, HBM, LR3, LRP7, OPPG, mKIAA4142
Chromosome Location
Chr.19: 3584825 - 3686564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008513.3 NP_032539.2 20 - 21 4467 64
GenBank mRNA AF064984.1 - 20 - 21 4457 64
AF077847.1 - 20 - 21 4345 64
AK090382.1 - 11 - 12 65
AK147068.1 - 20 - 21 4460 64
AK147231.1 - 20 - 21 8207 64
AK147330.1 - 20 - 21 8195 64
AK220186.1 - 20 - 21 4420 64
BC011374.1 - 20 - 21 4449 64
BC094610.1 - 20 - 21 4454 64

View Full Product Details