The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008866.2 | NP_032892.1 | 6 - 7 | 381 | 119 |
GenBank mRNA | AK002674.1 | - | 6 - 7 | 385 | 119 |
AK050549.1 | - | 6 - 7 | 358 | 119 | |
AK146874.1 | - | 6 - 7 | 396 | 119 | |
AK167231.1 | - | - | 403 | 119 | |
BC013536.1 | - | 6 - 7 | 381 | 119 | |
BC052848.1 | - | 6 - 7 | 422 | 119 | |
CT010201.1 | - | 6 - 7 | 360 | 119 | |
U89352.1 | - | 6 - 7 | 360 | 119 |