Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thy1
Entrez Gene ID
21838
Gene Name
thymus cell antigen 1, theta
Gene Aliases
CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2
Chromosome Location
Chr.9: 44043384 - 44048579 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009382.3 NP_033408.1 3 - 4 499 84
GenBank mRNA BC054436.1 - 2 - 3 410 84

View Full Product Details