Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uba1
Entrez Gene ID
22201
Gene Name
ubiquitin-like modifier activating enzyme 1
Gene Aliases
A1S9, Sbx, Ube-1, Ube1x
Chromosome Location
Chr.X: 20658302 - 20683179 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136085.2 NP_001129557.1 9 - 10 1142 73
NM_001276316.1 NP_001263245.1 10 - 11 1234 73
NM_001276317.1 NP_001263246.1 9 - 10 1137 73
NM_009457.4 NP_033483.2 9 - 10 1137 73
GenBank mRNA AK081808.1 - 9 - 10 1108 73
AK088057.1 - 9 - 10 1019 73
AK088528.1 - 9 - 10 1019 73
AK143416.1 - 9 - 10 1043 73
AK171667.1 - 9 - 10 1108 73
AK205753.1 - 2 - 3 174 73
AK219868.1 - 2 - 3 226 73
AK219938.1 - 2 - 3 226 73
BC058630.1 - 9 - 10 1083 73
BC138200.1 - 9 - 10 1000 73
BC145984.1 - 9 - 10 1001 73
D10576.1 - 9 - 10 1003 73

View Full Product Details