Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vamp2
Entrez Gene ID
22318
Gene Name
vesicle-associated membrane protein 2
Gene Aliases
Syb-2, Syb2, sybII
Chromosome Location
Chr.11: 69088528 - 69092381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009497.3 NP_033523.1 3 - 4 375 66
GenBank mRNA AF007168.1 - 2 - 3 277 66
AK090178.1 - 3 - 4 394 66
BC039745.1 - 3 - 4 356 66
BC055105.1 - 3 - 4 375 66
U60150.1 - 3 - 4 337 66

Gene Symbol
Ubr3
Entrez Gene ID
68795
Gene Name
ubiquitin protein ligase E3 component n-recognin 3
Gene Aliases
1110059H15Rik, 4833421P10Rik, A130030D10Rik, AA409735, AA414972, AA422631, AI646861, AU016126, Zfp650, Znf650
Chromosome Location
Chr.2: 69896969 - 70024010 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK035617.1 - - 134 66

View Full Product Details