Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xrcc1
Entrez Gene ID
22594
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 1
Gene Aliases
Xrcc-1
Chromosome Location
Chr.7: 24546300 - 24573438 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009532.4 NP_033558.3 16 - 17 1947 55
GenBank mRNA AK155376.1 - 16 - 17 1924 55
AK165569.1 - 16 - 17 1902 55
AK168334.1 - 16 - 17 1897 55
AK168727.1 - 16 - 17 1901 55
AU020161.1 - 1 - 2 55
BC055900.1 - 16 - 17 1923 55
BC085281.1 - 18 - 19 2329 55
U02887.1 - 16 - 17 1911 55

View Full Product Details