The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009541.2 | NP_033567.2 | 2 - 3 | 218 | 96 |
GenBank mRNA | AK051971.1 | - | 2 - 3 | 208 | 96 |
AK134251.1 | - | 2 - 3 | 178 | 96 | |
BC060055.1 | - | 2 - 3 | 192 | 96 | |
BC145312.1 | - | 1 - 2 | 91 | 96 | |
BC145313.1 | - | 1 - 2 | 91 | 96 | |
BC145821.1 | - | 1 - 2 | 91 | 96 | |
BY246264.1 | - | 2 - 3 | 219 | 96 | |
U14556.1 | - | 2 - 3 | 216 | 96 | |
U22396.1 | - | 2 - 3 | 206 | 96 |