Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb17
Entrez Gene ID
22642
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
AA589413, Lp-1, Miz1, Zfp100, mZ13
Chromosome Location
Chr.4: 141444659 - 141467937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009541.2 NP_033567.2 3 - 4 422 74
GenBank mRNA AK051971.1 - 3 - 4 412 74
AK134251.1 - 3 - 4 382 74
BC060055.1 - 3 - 4 396 74
BC145312.1 - 2 - 3 295 74
BC145313.1 - 2 - 3 295 74
BC145821.1 - 2 - 3 295 74
U14556.1 - 3 - 4 420 74
U22396.1 - 3 - 4 410 74

View Full Product Details