Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb17
Entrez Gene ID
22642
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
AA589413, Lp-1, Miz1, Zfp100, mZ13
Chromosome Location
Chr.4: 141444659 - 141467937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009541.2 NP_033567.2 8 - 9 1269 59
GenBank mRNA AK051971.1 - 7 - 8 1340 59
AK134251.1 - 8 - 9 1229 59
BC060055.1 - 7 - 8 1324 59
BC145312.1 - 7 - 8 1142 59
BC145313.1 - 7 - 8 1135 59
BC145821.1 - 7 - 8 1142 59
U14556.1 - 8 - 9 1267 59
U22396.1 - 8 - 9 1257 59

View Full Product Details