Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp92
Entrez Gene ID
22754
Gene Name
zinc finger protein 92
Gene Aliases
-
Chromosome Location
Chr.X: 73410978 - 73429813 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009566.4 NP_033592.2 2 - 3 130 65
GenBank mRNA AK141522.1 - 2 - 3 131 65
BC096436.1 - 1 - 2 181 65
U47104.1 - 2 - 3 150 65

View Full Product Details