The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009569.3 | NP_033595.1 | 9 - 10 | 1569 | 91 |
GenBank mRNA | AF006492.1 | - | 9 - 10 | 1573 | 91 |