The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009572.4 | NP_033598.2 | 2 - 3 | 460 | 110 |
GenBank mRNA | AK051410.1 | - | 2 - 3 | 449 | 110 |
AK053191.1 | - | 2 - 3 | 170 | 110 | |
AK220508.1 | - | 2 - 3 | 421 | 110 | |
BB656632.1 | - | 2 - 3 | 441 | 110 | |
Z54200.1 | - | 2 - 3 | 171 | 110 |