Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dct
Entrez Gene ID
13190
Gene Name
dopachrome tautomerase
Gene Aliases
DT, TRP-2, TRP2, Tyrp-2, Tyrp2, slaty, slt
Chromosome Location
Chr.14: 118012790 - 118052323 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010024.3 NP_034154.2 4 - 5 1325 65
GenBank mRNA AK033040.1 - 4 - 5 1324 65
BC067064.1 - 4 - 5 1159 65
BC082330.1 - 4 - 5 960 65
EU554632.1 - 4 - 5 857 65
X63349.1 - 4 - 5 1261 65

View Full Product Details