The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198872.1 | NP_001185801.1 | 17 - 18 | 1881 | 99 |
NM_001198873.1 | NP_001185802.1 | 17 - 18 | 1866 | 99 | |
NM_001198874.1 | NP_001185803.1 | 15 - 16 | 1812 | 99 | |
NM_001198875.1 | NP_001185804.1 | 15 - 16 | 1770 | 99 | |
NM_001198876.1 | NP_001185805.1 | 14 - 15 | 1752 | 99 | |
NM_001198877.1 | NP_001185806.1 | 17 - 18 | 1870 | 99 | |
NM_001198878.1 | NP_001185807.1 | 17 - 18 | 1855 | 99 | |
NM_010064.4 | NP_034194.1 | 14 - 15 | 1741 | 99 | |
GenBank mRNA | AF063231.1 | - | 14 - 15 | 1680 | 99 |
AK088015.1 | - | 14 - 15 | 1712 | 99 | |
AK141837.1 | - | 15 - 16 | 1780 | 99 | |
AK150173.1 | - | 14 - 15 | 1702 | 99 | |
AK164322.1 | - | 15 - 16 | 1736 | 99 | |
AK164437.1 | - | 14 - 15 | 1676 | 99 | |
AK168730.1 | - | 14 - 15 | 1716 | 99 | |
BC139374.1 | - | 14 - 15 | 1636 | 99 | |
BC139375.1 | - | 14 - 15 | 1636 | 99 | |
GU992208.1 | - | 17 - 18 | 1772 | 99 | |
GU992209.1 | - | 17 - 18 | 1757 | 99 | |
GU992210.1 | - | 15 - 16 | 1661 | 99 | |
GU992211.1 | - | 17 - 18 | 1810 | 99 | |
GU992212.1 | - | 17 - 18 | 1795 | 99 |