Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gfra3
Entrez Gene ID
14587
Gene Name
glial cell line derived neurotrophic factor family receptor alpha 3
Gene Aliases
GFRalpha3, Y15110
Chromosome Location
Chr.18: 34689903 - 34720387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010280.3 NP_034410.3 7 - 8 1269 73
GenBank mRNA AB008833.1 - 7 - 8 1147 73
AF020305.1 - 7 - 8 1102 73
AF036163.1 - 7 - 8 1163 73
AF041842.1 - 7 - 8 1152 73
AF051766.1 - 7 - 8 1164 73
AK046542.1 - 7 - 8 1196 73
BC066202.1 - 7 - 8 1269 73
Y15110.1 - 7 - 8 1183 73

View Full Product Details