The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010304.3 | NP_034434.1 | 1 - 2 | 419 | 58 |
GenBank mRNA | BC005439.1 | - | 1 - 2 | 419 | 58 |
BC011098.1 | - | 1 - 2 | 169 | 58 | |
M80632.1 | - | 1 - 2 | 190 | 58 |