The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001999.1 | NP_001001999.1 | 1 - 1 | 1509 | 57 |
NM_010327.2 | NP_034457.1 | 2 - 2 | 1271 | 57 | |
GenBank mRNA | BC053108.1 | - | 1 - 1 | 1509 | 57 |
BC138156.1 | - | 1 - 1 | 1463 | 57 | |
BC145160.1 | - | 1 - 1 | 1463 | 57 |