Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grb7
Entrez Gene ID
14786
Gene Name
growth factor receptor bound protein 7
Gene Aliases
mKIAA4028
Chromosome Location
Chr.11: 98446834 - 98455373 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010346.2 NP_034476.1 6 - 7 1083 89
GenBank mRNA AK057640.1 - 6 - 7 1076 89
AK145996.1 - 6 - 7 899 89
AK168438.1 - 6 - 7 867 89
AK220277.1 - 5 - 6 974 89
BC003295.1 - 6 - 7 826 89
BC081547.1 - 6 - 7 869 89
M94450.1 - 6 - 7 1105 89

View Full Product Details