Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2i
Entrez Gene ID
14886
Gene Name
general transcription factor II I
Gene Aliases
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location
Chr.5: 134237832 - 134314760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 14 - 15 1505 69
NM_001080747.2 NP_001074216.1 13 - 14 1442 69
NM_001080748.2 NP_001074217.1 12 - 13 1385 69
NM_001080749.2 NP_001074218.1 12 - 13 1385 69
NM_010365.4 NP_034495.2 13 - 14 1448 69
GenBank mRNA AF017085.1 - 14 - 15 1149 69
AF043219.1 - 12 - 13 1069 69
AF043220.1 - 11 - 12 1006 69
AK145698.1 - 4 - 5 334 69
AK147201.1 - 12 - 13 1343 69
AY030290.1 - 13 - 14 1135 69
AY030291.1 - 13 - 14 1141 69
AY030292.1 - 12 - 13 1078 69
AY030293.1 - 14 - 15 1198 69
BC047387.1 - 12 - 13 1033 69
BC053044.1 - 14 - 15 1474 69

View Full Product Details