The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136067.2 | NP_001129539.1 | 9 - 10 | 1292 | 71 |
NM_001161421.1 | NP_001154893.1 | 9 - 10 | 1289 | 71 | |
NM_001161422.1 | NP_001154894.1 | 8 - 9 | 1110 | 71 | |
NM_001161423.1 | NP_001154895.1 | 8 - 9 | 1162 | 71 | |
NM_001161424.1 | NP_001154896.1 | 8 - 9 | 1159 | 71 | |
NM_010547.2 | NP_034677.2 | 8 - 9 | 1188 | 71 | |
NM_178590.4 | NP_848705.1 | 8 - 9 | 1185 | 71 | |
GenBank mRNA | AF069542.1 | - | 7 - 8 | 1047 | 71 |
AF513109.1 | - | 9 - 10 | 1203 | 71 | |
AK037020.1 | - | 8 - 9 | 1164 | 71 | |
AK154095.1 | - | 8 - 9 | 1174 | 71 | |
AY112937.1 | - | 9 - 10 | 1292 | 71 | |
BC021431.1 | - | 9 - 10 | 1279 | 71 |