The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010673.2 | NP_034803.2 | 1 - 1 | 448 | 117 |
GenBank mRNA | BC116356.1 | - | 1 - 1 | 447 | 117 |
BC116357.1 | - | 1 - 1 | 447 | 117 | |
BC128485.1 | - | 1 - 1 | 447 | 117 | |
BC128486.1 | - | 1 - 1 | 447 | 117 | |
D89902.1 | - | 2 - 2 | 460 | 117 |