The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159577.1 | NP_001153049.1 | 4 - 5 | 985 | 71 |
NM_001159578.1 | NP_001153050.1 | 5 - 6 | 1130 | 71 | |
NM_001159579.1 | NP_001153051.1 | 3 - 4 | 761 | 71 | |
NM_010727.4 | NP_034857.3 | 3 - 4 | 761 | 71 | |
GenBank mRNA | AF034745.1 | - | 4 - 5 | 985 | 71 |
AF034746.1 | - | 3 - 4 | 689 | 71 | |
AK161194.1 | - | 5 - 6 | 1116 | 71 | |
BC040367.1 | - | 3 - 4 | 713 | 71 | |
BC139771.1 | - | 3 - 4 | 689 | 71 | |
KJ418422.1 | - | 3 - 4 | 459 | 71 |