The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159577.1 | NP_001153049.1 | 9 - 10 | 2087 | 85 |
NM_001159578.1 | NP_001153050.1 | 10 - 11 | 2232 | 85 | |
NM_001159579.1 | NP_001153051.1 | 8 - 9 | 1863 | 85 | |
NM_001159580.1 | NP_001153052.1 | 8 - 9 | 1833 | 85 | |
NM_010727.4 | NP_034857.3 | 8 - 9 | 1863 | 85 | |
GenBank mRNA | AF034745.1 | - | 9 - 10 | 2087 | 85 |
AF034746.1 | - | 8 - 9 | 1791 | 85 | |
AK044127.1 | - | 8 - 9 | 1816 | 85 | |
AK138326.1 | - | 8 - 9 | 1834 | 85 | |
AK161194.1 | - | 10 - 11 | 2218 | 85 | |
BC139771.1 | - | 8 - 9 | 1791 | 85 | |
KJ418422.1 | - | 8 - 9 | 1561 | 85 |