The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011153.3 | NP_035283.1 | 2 - 3 | 218 | 92 |
GenBank mRNA | AF071562.1 | - | 2 - 3 | 216 | 92 |
AF500906.1 | - | 2 - 3 | 228 | 92 | |
AK078335.1 | - | 2 - 3 | 235 | 92 | |
BC026822.1 | - | 2 - 3 | 218 | 92 |