Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc23a1
Entrez Gene ID
20522
Gene Name
solute carrier family 23 (nucleobase transporters), member 1
Gene Aliases
D18Ucla2, SVCT1, Slc23a2, YSPL3
Chromosome Location
Chr.18: 35604041 - 35630340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011397.4 NP_035527.3 8 - 9 1035 89
GenBank mRNA AF058318.1 - 8 - 9 945 89
AK085499.1 - 8 - 9 950 89
AK143959.1 - 8 - 9 1036 89
AK165439.1 - 8 - 9 971 89
BC013528.1 - 8 - 9 964 89

View Full Product Details