Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfrsf17
Entrez Gene ID
21935
Gene Name
tumor necrosis factor receptor superfamily, member 17
Gene Aliases
BCM, BCMA, Tnfrsf13, Tnfrsf13a
Chromosome Location
Chr.16: 11313174 - 11320074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011608.1 NP_035738.1 1 - 2 257 61
GenBank mRNA AF061505.1 - 1 - 2 257 61
AK020247.1 - 1 - 2 255 61
BC027519.1 - 1 - 2 128 61

View Full Product Details