Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tpbg
Entrez Gene ID
21983
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, AW495680, WAIF1
Chromosome Location
Chr.9: 85842380 - 85847055 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164792.1 NP_001158264.1 2 - 2 790 94
NM_011627.4 NP_035757.2 2 - 2 899 94
GenBank mRNA AK051162.1 - 2 - 2 870 94
AK143519.1 - 2 - 2 729 94
AK165553.1 - 2 - 2 780 94
AK165597.1 - 2 - 2 902 94
BC058198.1 - 2 - 2 758 94

View Full Product Details