The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286122.1 | NP_001273051.1 | 9 - 10 | 1308 | 106 |
NM_011632.3 | NP_035762.2 | 11 - 12 | 1494 | 106 | |
GenBank mRNA | AK147304.1 | - | 9 - 10 | 1236 | 106 |
AK147362.1 | - | 9 - 10 | 1309 | 106 | |
AK148231.1 | - | 5 - 6 | 649 | 106 | |
BC137634.1 | - | 10 - 11 | 1209 | 106 | |
BC137635.1 | - | 10 - 11 | 1209 | 106 | |
U21050.1 | - | 11 - 12 | 1313 | 106 |