The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011633.2 | NP_035763.2 | - | 1448 | 75 |
GenBank mRNA | AK144694.1 | - | - | 1399 | 75 |
BC012702.1 | - | - | 1402 | 75 | |
D78141.1 | - | - | 1425 | 75 | |
D83528.1 | - | - | 1290 | 75 |