Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tulp3
Entrez Gene ID
22158
Gene Name
tubby-like protein 3
Gene Aliases
2310022L06Rik, AI316887
Chromosome Location
Chr.6: 128321161 - 128355872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011657.2 NP_035787.1 1 - 2 113 83
GenBank mRNA AB016963.1 - 1 - 2 56 83
AF045582.1 - 1 - 2 66 83
AK035363.1 - 1 - 2 78 83
AK142527.1 - 1 - 2 103 83
BY347356.1 - 1 - 2 116 83

View Full Product Details