Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tulp3
Entrez Gene ID
22158
Gene Name
tubby-like protein 3
Gene Aliases
2310022L06Rik, AI316887
Chromosome Location
Chr.6: 128321161 - 128355872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011657.2 NP_035787.1 8 - 9 1045 108
GenBank mRNA AB016963.1 - 8 - 9 988 108
AF045582.1 - 8 - 9 998 108
BC060068.1 - 8 - 9 975 108

View Full Product Details