The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011667.2 | NP_035797.1 | 12 - 13 | 1440 | 67 |
GenBank mRNA | AF150963.1 | - | 12 - 13 | 1440 | 67 |
AK030031.1 | - | 12 - 13 | 1439 | 67 |