Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ugcg
Entrez Gene ID
22234
Gene Name
UDP-glucose ceramide glucosyltransferase
Gene Aliases
AU043821, C80537, Epcs21, GlcT-1, Ugcgl
Chromosome Location
Chr.4: 59189204 - 59222833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011673.3 NP_035803.1 5 - 6 652 100
GenBank mRNA AK138848.1 - 5 - 6 99
BC050828.1 - 5 - 6 652 100
D89866.1 - 5 - 6 743 100

View Full Product Details