The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011673.3 | NP_035803.1 | 6 - 7 | 827 | 96 |
GenBank mRNA | AK138848.1 | - | 6 - 7 | 842 | 96 |
BC050828.1 | - | 6 - 7 | 827 | 96 | |
D89866.1 | - | 6 - 7 | 918 | 96 |