Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vnn1
Entrez Gene ID
22361
Gene Name
vanin 1
Gene Aliases
V-1
Chromosome Location
Chr.10: 23894688 - 23905344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011704.3 NP_035834.2 4 - 5 1016 82
GenBank mRNA AJ132098.1 - 4 - 5 849 82
AK002773.1 - 4 - 5 848 82
AK145984.1 - 4 - 5 922 82
AK149485.1 - 2 - 3 397 82
AK167427.1 - 4 - 5 848 82
BC019203.1 - 4 - 5 842 82

View Full Product Details